missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Orion™ Ion Specific Electrode Kits

Ensure correct measurement by using high quality electrodes and reagents for ion specific measurement. Ordering is easy! All the consumables you need to measure fluoride or ammonia in water and wastewater are available with one part number. Now with NEW 0.5 ppm and 5.0 ppm Fluoride Standards with TISAB II for low level fluoride analysis

Brand:  Thermo Scientific™ 9609BNLSLN

704.65 EUR valid until 2024-12-31
Use promo code "21658" to get your promotional price.


 View more versions of this product

Product Code. 15839656

  • 859.28 € / Each
Estimated Shipment: 13-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Perform quick and accurate measurements by using high quality Thermo Scientific™ Orion™ Ion Specific Electrodes and reagents for ion specific measurement. Ordering is easy! We offer a comprehensive portfolio of ion selective electrode calibration standards, ionic strength adjusters, reagents, and fill solutions for your ISE measurement needs. All the consumables you need to measure fluoride or ammonia in water and wastewater are available with one part number. Now with NEW 0.5 ppm and 5.0 ppm Fluoride Standards with TISAB II for low level fluoride analysis.

Fluoride ISE Kit


  • Orion Combination Fluoride ISE features the Thermo™ Scientific™ Orion™ Sure-Flow™ junction which contributes to highly stable and precise fluoride measurements while simplifying electrode maintenance.
  • 0.5, 1.0, 2.0, 5.0 and 10.0 ppm Fluoride Standards are pre-mixed with total ionic strength adjustor for your convenience. No dilution and no addition of ionic strength adjustor needed.
  • Total Ionic Strength Adjustor (TISAB II) included for addition to samples
Ammonia ISE Kit


  • This kit includes the Orion Standard Performance Ammonia ISE (Cat. No. 9512BNWP) and is ideal for mid to high ammonia level measurements.
  • The Ionic Strength Adjuster (ISA) (Cat. No. 951211) included in the kit, is formulated for samples containing metallic ions and has pH indicating blue dye added, so you know the pH is correct for analysis.
  • This kit includes the Orion Standard Performance Ammonia ISE (Cat. No. 9512BNWP) and is ideal for mid to high ammonia level measurements.
  • The Ionic Strength Adjuster (ISA) (Cat. No. 951211) included in the kit, is formulated for samples containing metallic ions and has pH indicating blue dye added, so you know the pH is correct for analysis.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Fluoride Analysis
Low Level Fluoride Ion Selective Electrode and Reagent Kit
14mm
Optimum Results A fill sol PN 90061
Sure-Flow
39.40 in.
1m
RANGES: 1 x 10E-6M to saturated
Electrode,Standards, ISA
9609BNWP
0-80°C
ISE Meter
Combination ISE and Reagent Kit
BNC, waterproof
0.55 in.
17mm
9609BNWP
4.75 in.
110mm
Epoxy
32-180°F
pellet
Fluoride ISE
Kit
25°C
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers