Learn More
Thermo Scientific™ Orion™ ISE Filling Solutions
Enhance the performance and extend the life of your ion selective electrodes with these ISE filling solutions.
86.63 € - 160.80 €
Specifications
For Use With (Equipment) | Thermo Scientific Orion Ion Selective Electrodes |
---|---|
Type | Ion Selective Electrode Filling Solution |
Product Code | Brand | Description | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | ||||||
12084502
|
Thermo Scientific™
900010 |
Fill Solution for Sodium ISE |
153.95 €
Pack of 5 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
12048882
|
Thermo Scientific™
900012 |
Fill Solution for Sodium ISE with Low Sodium Sample Levels |
150.75 €
Pack of 5 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
11727797
|
Thermo Scientific™
900017 |
Fill Solution for Chloride ISE |
107.54 €
Pack of 5 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
11762787
|
Thermo Scientific™
900018-WA |
Fill Solution For Ammonium ISE |
148.74 €
Pack of 5 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
12074502
|
Thermo Scientific™
900046 |
Optimum Results F Fill Solution for Nitrate and Nitrite ISE |
122.61 €
Pack of 5 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
12054502
|
Thermo Scientific™
900061 |
Optimum Results A Fill Solution for Cadmium, Calcium, Fluoride and Sulfide ISE |
124.82 €
Pack of 5 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
11854333
|
Thermo Scientific™
900062 |
Optimum Results B Fill Solution for Chloride, Cyanide, Lead and Silver/Sulfide ISE |
127.51 €
Pack of 5 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
11956079
|
Thermo Scientific™
900063 |
Optimum Results D Fill Solution for Bromide, Cupric and Iodide ISE |
120.60 €
Pack of 5 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
11737797
|
Thermo Scientific™
900065 |
Optimum Results E Fill Solution for Potassium ISE |
127.64 €
Pack of 5 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
11747797
|
Thermo Scientific™
900067 |
Optimum Results C Fill Solution for Silver ISE |
160.80 €
Pack of 5 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
11717677
|
Thermo Scientific™
950202 |
Fill Solution for Carbon Dioxide ISE |
101.51 €
60mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
10675261
|
Thermo Scientific™
951202 |
Fill Solution For Ammonia ISE | N/A | N/A | N/A | |||||
12024502
|
Thermo Scientific™
951202 |
Fill Solution For Ammonia ISE |
86.63 €
60mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
10368883
|
Thermo Scientific™
951209 |
Fill Solution For Ammonia High Performance ISE | N/A | N/A | N/A | |||||
12871521
|
Thermo Scientific™
951209 |
Fill Solution For Ammonia High Performance ISE |
86.63 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
Enhance the performance and extend the life of your ion selective electrodes with Thermo Scientific™ Orion™ ISE Filling Solutions. These solutions are specially formulated to work with the recommended ion selective electrodes and prevent electrode interferences from filling solution.
Features:- Filling/maintenance solutions are for use with Thermo Scientific™ Orion™ Ion Selective Electrodes (ISE)
- Optimum Result™ ISE filling solutions are specially designed for the Ionplus™ series ion selective electrodes
Ordering Information:
- Packaged as set of five 2 oz. (5 x 60mL) bottles, except Cat. No. 951209, 951202 and 950202 packaged as one 2 oz. (60mL) bottle and Cat. No. 954602 packaged as one 1.6 oz. (50mL) bottle
Note:
The following part number has been changed due to a recent system upgrade:
Old Part Number: 900018
New Part Number: 900018-WA
Please use the new part number when placing an order or requesting a quote. The old and new part numbers are interchangeable on Certificate of Analysis and Material Safety Data Sheet (MSDS) files.
Warranty: Out of box
Specifications
Thermo Scientific Orion Ion Selective Electrodes | |
Ion Selective Electrode Filling Solution |
Safety and Handling
- Sodium Reference Electrode Filling Solution 2M NH4Cl
Signal Word
- Warning
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 3
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H319-Causes serious eye irritation.
- H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
- P202-Do not handle until all safety precautions have been read and understood.
- P273-Avoid release to the environment.
Supplemental information
- MIXTURE LIST-Contains : ammonium chloride
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.