Learn More
Thermo Scientific™ Orion Lab Star Bench Meter Accessories
Orion Lab Star series meter accessories support your Orion Lab Star PH111 pH meter, Orion Lab Star EC112 conductivity meter or Orion Lab Star DO113 DO meter.
53.87 € - 106.53 €
Specifications
Type Accessory
Compatibility Orion Lab Star PH111 pH meter, Orion Lab Star EC112 conductivity meter and Orion Lab Star DO113 DO meter
View More Specs
Products 4
Product Code Brand Description Certifications/Compliance Price Quantity & Availability  
Product Code Brand Description Certifications/Compliance Price Quantity & Availability  
17701849
View Documents
Thermo Scientific™
LSTAR-ARM
Orion Lab Star series electrode stand with meter-attachable bracket None
106.53 €
Each
In Stock
to see stock.
Add to basket
17711849
View Documents
Thermo Scientific™
LSTAR-CVR
Orion Lab Star series meter dust cover None
53.87 €
Each
In Stock
to see stock.
Add to basket
17711749
View Documents
Thermo Scientific™
LSTAR-PWR
Orion Lab Star series 100-240 V 50/60 Hz universal power adapter with twist-and-lock connector CE, CSA, FCC Class A limits
53.87 €
Each
Estimated Shipment: 20-02-2025
to see stock.
Add to basket
17721849
View Documents
Thermo Scientific™
LSTAR-USB
Orion Lab Star series USB computer cable None
53.87 €
Each
In Stock
to see stock.
Add to basket
Description
Description
Select Orion Lab Star series meter accessories to support your Orion Lab Star PH111 pH/mV meter, Orion Lab Star EC112 conductivity meter or Orion Lab Star DO113 dissolved oxygen meter. Utilize the meter dust cover accessory for extended meter storage. Select a replacement meter-attached electrode holder, universal power adapter or computer cable when needed. The Orion Lab Star series pH meter, conductivity meter and DO meter are ideal for simplified, budget-friendly lab testing and data recording.
Orion Lab Star Series Meter Accessories Features
  • LSTAR-ARM Orion Lab Star series meter-attached electrode stand allows you to replace your electrode stand as needed and is compatible with the Orion Lab Star PH111 pH meter, Orion Lab Star EC112 conductivity meter and Orion Lab Star DO113 DO meter
  • LSTAR-PWR Orion Lab Star series universal power supply, 100-240V 50/60 Hz, allows you to replace your power adapter as needed and is compatible with the Orion Lab Star PH111 pH meter, Orion Lab Star EC112 conductivity meter and Orion Lab Star DO113 DO meter
  • LSTAR-USB Orion Lab Star series USB computer cable allows you to replace your computer cable as needed and is compatible with the Orion Lab Star PH111 pH meter, Orion Lab Star EC112 conductivity meter and Orion Lab Star DO113 DO meter
  • LSTAR-CVR Orion Lab Star series meter dust cover protects your meter from dust and debris while in storage and is compatible with the Orion Lab Star PH111 pH meter, Orion Lab Star EC112 conductivity meter and Orion Lab Star DO113 DO meter
Additional Accessories Compatible with Orion Lab Star Series Meters
  • STARA-106 ink-ribbon compact printer, 100-240V 50/60 Hz
Specifications
Specifications
Accessory
Orion Lab Star PH111 pH meter, Orion Lab Star EC112 conductivity meter and Orion Lab Star DO113 DO meter