missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Orion™ Nitrate Electrodes

Measure nitrate in aqueous solutions simply, accurately and economically with these electrodes.

Brand:  Thermo Scientific™ 9307BNWP

728.45 EUR valid until 2024-12-31
Use promo code "21658" to get your promotional price.


 View more versions of this product

Product Code. 10049140

  • 882.39 € / Each
Estimated Shipment: 06-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Measure nitrate in aqueous solutions simply, accurately and economically with Thermo Scientific™ Orion™ Nitrate Electrodes.

9707BNWP is a combination electrode with a Sure-Flow™ junction. When conventional electrode junctions clog, the Sure-Flow junction can be opened and flushed clean simply by pressing the cap. To be used with module 970701.

Nitrate Half-Cell Electrode 9307BNWP (comes with half cell module 930702) is to be used with the Orion 900200 sure flow double junction reference electrode.

  • Calibration standards catalog number: 920706 — 0.1M NaNO3; 920707 — 1000ppm N; 930707 — 100ppm N
  • Required ISA catalog number: 930711 or 930710/nitrate ISS
TRUSTED_SUSTAINABILITY
Specifications

Specifications

BNC
drift-free potentials
ISE Meters
Half-Cell Nitrate Electrode with Waterproof BNC Connector
Free-flowing Sure-Flow™ junction assures stable
Combination Ion Selective Electrode
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers