Learn More
Thermo Scientific™ Orion™ Nitrate Electrodes
Measure nitrate in aqueous solutions simply, accurately and economically with these electrodes.
Brand: Thermo Scientific™ 9707BNWP
824.50 EUR valid until 2024-12-31
Use promo code "21658" to get your promotional price.
Description
Measure nitrate in aqueous solutions simply, accurately and economically with Thermo Scientific™ Orion™ Nitrate Electrodes.
9707BNWP is a combination electrode with a Sure-Flow™ junction. When conventional electrode junctions clog, the Sure-Flow junction can be opened and flushed clean simply by pressing the cap. To be used with module 970701.
Nitrate Half-Cell Electrode 9307BNWP (comes with half cell module 930702) is to be used with the Orion 900200 sure flow double junction reference electrode.
- Calibration standards catalog number: 920706 — 0.1M NaNO3; 920707 — 1000ppm N; 930707 — 100ppm N
- Required ISA catalog number: 930711 or 930710/nitrate ISS
Specifications
BNC | |
drift-free potentials | |
ISE Meters |
Combination Nitrate Electrode with Waterproof BNC Connector | |
Free-flowing Sure-Flow™ junction assures stable | |
Combination Ion Selective Electrode |