Learn More
Thermo Scientific™ Orion™ ORP Standards for Redox/ORP Electrodes
Measure ORP simply and accurately with these standards, stable for up to a year (unopened).
Brand: Thermo Scientific™ 967961
View more versions of this product
Description
Measure ORP simply and accurately with Thermo Scientific™ Orion™ ORP Standards for Redox/ORP Electrodes. Orion OPR standards are stable for up to a year (unopened) and can be used directly from the bottle with no mixing or diluting.
- Included in the new ASTM ORP method
- Used as relative mV standard to calibrate the offset of sample
- +220mV vs. Ag/AgCl using 4M KCl saturated with AgCl filling solution (Part No. 900011), +420mV vs. Standard Hydrogen Electrode (EH)
Specifications
ORP Standard | |
5 x 60 mL | |
Gel-filled ORP/ATC Triode™ electrodes (9179BNMD and 9179BN), Refillable ORP/ATC Triode electrodes (9180BNMD and 9180BN), Glass-bodied ORP electrodes, (9778BNWP and 9778SC), Sure-Flow™ ORP electrodes, (9678BNWP and 967800) |
Bottle | |
Redox/ORP Electrode Standard |
Safety and Handling
- ORP Standard
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 3
- Serious eye damage/eye irritation Category 2
- Reproductive toxicity Category 1B
- Skin corrosion/irritation Category 2
Hazard Statement
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H360-May damage fertility or the unborn child.
- H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
- P201-Obtain special instructions before use.
- P202-Do not handle until all safety precautions have been read and understood.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P321-Specific treatment (see supplemental first aid instructions on this label).
Supplemental information
- MIXTURE LIST-Contains : potassium hydroxide,boric acid
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.