missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Orion™ pH Buffer Cubitainers™

Product Code. p-4521266
Click to view available options
Quantity:
19 L
pH:
4.01
7
10.01
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 11742817

Brand: Thermo Scientific™ 9104CB

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Hold 5 gallons (19L) of pH buffer in color-coded Thermo Scientific™ Orion™ pH Buffer Cubitainers™ with easy-pour spigot.

Hold 5 gallons (19L) of pH buffer in color-coded Thermo Scientific™ Orion™ pH Buffer Cubitainers™ with easy-pour spigot.

  • Color coded: pH 4.01 buffer is pink, pH 7.00 buffer is yellow, pH 10.01 buffer is blue
  • Packaged with buffer/temperature tables for accurate calibrations and NIST™ traceability
  • Manufactured under ISO 9000 quality standards
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material pH Buffer, 5 gallon cubitainers
Boiling Point 100°C
Color Light Red
Description pH 4.01 Buffer, 5 gallon
For Use With (Application) Accurate, reproducible pH calibrations
For Use With (Equipment) pH Meter and Electrode
pH 4.01
Physical Form Liquid
Quantity 19 L
Formulation Water, Potassium Hydroxide, Amaranth Red No. 27, Potassium Hydrogen Phthalate
Packaging Cubitainer
Traceability to NIST Yes
Solubility Information Soluble
Show More Show Less

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.