Learn More
Thermo Scientific™ Orion™ pH Electrode Storage Solutions
Restore proper electrode performance and prolong electrode life using these pH electrode storage solutions.
Brand: Thermo Scientific™ 9100CB
Description
Restore proper electrode performance and prolong electrode life using Thermo Scientific™ Orion™ pH Electrode Storage Solutions in ready to use bottles.
Storage Solutions:
- Storage solution ensures that the pH electrode works properly after prolonged storage, overnight storage and between samples
Warranty: Out of box

Specifications
pH Electrode Storage Solution, 5 gallon (19L) Cubitainer | |
pH Electrode Storage Solution | |
Cubitainer |
pH Electrode | |
Ensures that the pH electrode works properly after prolonged storage, overnight storage and between samples | |
19 L |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.