Learn More
Thermo Scientific™ Orion™ pH Electrode Storage Solutions
Restore proper electrode performance and prolong electrode life using these pH electrode storage solutions.
Brand: Thermo Scientific™ 810001
84.96 EUR valid until 2025-04-30
Use promo code "24155" to get your promotional price.
Description
Restore proper electrode performance and prolong electrode life using Thermo Scientific™ Orion™ pH Electrode Storage Solutions in ready to use bottles.
Storage Solutions:
- Storage solution ensures that the pH electrode works properly after prolonged storage, overnight storage and between samples
Warranty: Out of box

Specifications
ROSS pH Electrode Storage Solution, 1 pint (475mL) Bottle | |
pH Electrode Storage Solution | |
Bottle |
pH Electrode | |
Storage | |
475 mL |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.