missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Orion™ pH Electrode Cleaning Solutions

Restore proper electrode performance and prolong electrode life using these pH electrode cleaning solutions.

Brand:  Thermo Scientific™ 510015

 View more versions of this product

Product Code. 11722737

  • 35.38 € / 60mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Restore proper electrode performance and prolong electrode life using Thermo Scientific™ Orion™ pH Electrode Cleaning Solutions in ready to use bottles.

Cleaning Solutions:

 

  • Solutions are identified by the letters A, B, C, and D; each letter designates a unique formulation
  • Cleaning solution A is for removing protein contaminants, cleaning solution B is for remov
    TRUSTED_SUSTAINABILITY
Specifications

Specifications

2 oz. (60mL) Bottle of No Cal Stain Lifter Solution
pH Electrode Cleaning Solution
Solutions restore proper electrode performance and prolong the useful life of the electrode
60 mL
pH Electrode
Cleaning
Bottle
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberBV1 Certificate TypeCertificate of Analysis Date16/06/2023 Catalog Number510015
Lot NumberBV1 Certificate TypeCertificate of Analysis Date15/06/2023 Catalog Number510015
Lot NumberBV1 Certificate TypeCertificate of Analysis Date09/06/2023 Catalog Number510015
Lot NumberBV1 Certificate TypeCertificate of Analysis Date09/06/2023 Catalog Number510015
Lot NumberXQ1 Certificate TypeCertificate of Analysis Date09/10/2019 Catalog Number510015
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Orion™ pH Electrode Cleaning Solutions >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.