Documents
Product Information
Manuals
- 1
- 2
Achieve easier and cleaner measurements with pre-measured and ready-to-use powder chemistry packs.
Brand: Thermo Scientific™ AC4P72
36.47 EUR valid until 2025-07-13
Use promo code "24135" to get your promotional price.
Use Thermo Scientific™ Orion™ AQUAfast Powder Pack Reagents with Orion AQUAfast colorimeters and Thermo Scientific Orion AquaMate Vis and UV-Vis spectrophotometers. Powder pack chemistries dissolve quickly in solution, are easy to mix and do not require a stir rod or crushing. Each powder pack is notched for simplified pouch opening in the lab or field. Powder pack reagents measure aluminum, ammonia, chlorine, copper, cyanide, DEHA, iron, manganese, molybdate, nitrite, phosphate, silica and sulfate.
AQUAfast chlorine (total) reagent powder packs, 100 tests | |
Drinking Water: Standard Methods 4500-Cl G / ATP EPA-approved Orion Method AC4P72; Wastewater: Standard Methods 4500-Cl G-2000 / ATP EPA-approved Orion Method AC4P72 | |
100 Chlorine Total-DPD Powder Packs | |
100 tests | |
AQUAfast 24 mm round vials (Cat. No. AC2V24) |
Chlorine (total) | |
Orion AquaMate Vis and UV-Vis Spectrophotometers, Orion AQUAfast AQ4000 Colorimeter, Orion AQUAfast AQ3700 Colorimeter | |
Powder Pack | |
0.02 to 2 mg/L Cl2 | |
DPD |
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
YB1C1127 | Certificate of Analysis | 08/05/2023 | AC4P72 |
XB1E1126_01Y469178 | Certificate of Analysis | 08/01/2023 | AC4P72 |
XA8B0826 | Certificate of Analysis | 24/06/2022 | AC4P72 |
XA6A0626 | Certificate of Analysis | 24/06/2022 | AC4P72 |
XA6A0626 | Certificate of Analysis | 15/02/2022 | AC4P72 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.