Learn More
Thermo Scientific™ Orion™ RDO Replacement Electrode Cap
Use the probe optical cap replacement with RDO probes.
Brand:  Thermo Scientific™ 087001
133.36 EUR valid until 2025-07-11
Use promo code "25721" to get your promotional price.
Product Code. 10436758
  • 147.38 € / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
Use the Thermo Scientific™ Orion™ RDO™ Probe Optical Cap Replacement with Orion RDO probes.
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Optical Cap
Orion RDO Probes 087010MD, 087020MD, 087030MD, 087050MD, 087100MD
Probe Optical Cap
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Thermo Scientific™ Orion™ RDO Replacement Electrode Cap > For Orion RDO Probes 087010MD, 087020MD, 087030MD, 087050MD, 087100MD
Do not use solvents or soaps to clean the RDO cap and do not rub the cap with abrasive material. Use of these materials will damage the RDO cap and void the warranty. Do not let salt or mineral deposits form on the RDO cap.