Learn More
Thermo Scientific™ Orion™ Redox/ORP/Temp Electrodes
Efficiently measure ORP and temperature simultaneously without a separate reference electrode in a single instance with these redox/ORP/temp electrodes.
Brand:  Thermo Scientific™ 9179BNMD
321.30 EUR valid until 2024-12-31
Use promo code "21658" to get your promotional price.
 View more versions of this product
Product Code. 11732857
  • 379.89 € / Each
Estimated Shipment: 13-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
Efficiently measure ORP and temperature simultaneously without a separate reference electrode in a single instance. Thermo Scientific™ Orion™ Redox/ORP/Temp Electrodes measure ORP and temperature using a platinum redox sensor, a silver/silver chloride internal reference system and a built-in thermistor which is adapted for Automatic Temperature Compensation (ATC).
Features:
  • Available in epoxy triode low maintenance and epoxy refillable triode construction
  • Built-in thermistor for Automatic Temperature Compensation (ATC)
  • Not designed for amperometric or constant current titrations, such as Karl Fischer analysis
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Epoxy Triode Low Maintenance
Epoxy
Orion Star Series Meters
Redox/ORP/Temp Electrode