missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Orion™ ROSS Ultra™ Glass Bodied pH/ATC Triode™

Accurately measure pH and temperature together in just one electrode, with a built-in temperature sensor.

Brand:  Thermo Scientific™ 8302BNURCA

 View more versions of this product

Product Code. 16690713

  • 441.20 € / Each
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Thermo Scientific™ Refillable Orion™ ROSS Ultra™ Triode™ 3-in-1 pH/Automatic Temperature Compensation Electrodes combine the precision and speed of response of the ROSS Ultra™ line with a built-in ATC. Best suited for use in strong acids/bases, pH > 12 or pH < 2, food & cosmetic emulsions, pure water and slurries. Refillable Orion™ ROSS Ultra™ Triodes™ come with a two year warranty.
 

Measure pH and Temperature with One Electrode

Complete your pH measurements faster. Our ROSS Ultra Triode pH electrodes offer more flexibility, accuracy and precision for your pH measurement needs. With built-in temperature sensors, these electrodes offer a convenient solution for your routine sample measurements. Compatible with Thermo Scientific™ Orion™ Versa Star™, Thermo Scientific™ Orion Star™ A, and Thermo Scientific™ Orion™ Dual Star™ meters, as well as Mettler Toledo™ pH meters.

Target Markets:
  • Pharmaceutical
  • Food Manufacturing
  • Academic/Government
  • Environmental
Applications:
  • Quality Control
  • Research
  • General Purpose
Warranty: 24 months from date of purchase by Buyer or 36 months from date of shipment from Seller, whichever is earlier
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Orion ROSS Ultra Glass Triode pH/ATC Combination Electrodes with a BNC and RCA Connector and 1 meter cable
Ceramic
0 to 14
pH/ATC Combination Electrode
Mettler Toledo Five and Seven series pH meters
7
0.01
120 mm
12 mm
BNC Waterproof, RCA
Chemical Resistant Glass Body
0°C to 100°C
ROSS electrode 3 M KCl fill solution (Cat. No. 810007)
ROSS
0.03 pH for buffers using automatic temperature compensation
2 Years
1 m
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates
View accessory products
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Orion™ ROSS Ultra™ Glass Bodied pH/ATC Triode™ > With BNC and RCA Connector; for use with Mettler Toledo Five and Seven series pH meters

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.