Learn More
Thermo Scientific™ Orion™ ROSS Ultra™ Refillable pH/ATC Triode™ Combination Electrodes for Orion Series Meters
Measure biological media, pharmaceuticals, foods and more with Thermo Scientific™ Orion™ ROSS Ultra™ Refillable pH/ATC Triode™ Combination Electrodes.
Brand: Thermo Scientific™ 8157BNURCA
354.35 EUR valid until 2024-12-15
Use promo code "21658" to get your promotional price.
Description
Measure biological media, pharmaceuticals, foods and more with Thermo Scientific™ Orion™ ROSS Ultra™ Refillable pH/ATC Triode™ Combination Electrodes. They are ideal for any application where the use of metallic reference electrodes would contaminate the samples. ROSS Ultra pH/ATC electrodes offer more flexibility, accuracy and precision for your pH measurement needs. With built-in temperature sensors, these electrodes offer a convenient solution for your routine sample measurements. They have great stability and drift and come with a two-year replacement warranty.
Measure pH and Temperature with One Electrode
Measure biological media, pharmaceuticals, foods and more with Thermo Scientific™ Orion™ ROSS Ultra™ Refillable pH/ATC Triode™ Combination Electrodes. They are ideal for any application where the use of metallic reference electrodes would contaminate the samples. ROSS Ultra electrodes offer the best temperature performance in repeated, varying temperatures of all other electrodes. They have great stability and drift and come with a two-year replacement warranty.
Maintain sample integrity, even in TRIS buffers; ROSS double junction design electrodes contain no silver, which can bind with biological samples. With built-in temperature sensors, these electrodes offer a convenient solution for your routine sample measurements.
Compatible with Thermo Scientific™ Orion™ Versa Star™, Thermo Scientific™ Orion Star™ A, and Thermo Scientific™ Orion™ Dual Star™ meters, as well as Mettler Toledo™ pH meters.
Target Markets:
- Pharmaceutical/Biopharmaceutical
- Food Manufacturing
- Academic/Government
- Environmental
Applications:
- Quality Control
- Research
- General Purpose
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Orion ROSS Ultra Refillable pH/ATC Triode Combination Electrodes with a BNC and RCA Connector and 1 meter cable | |
Glass Fiber | |
0 to 14 | |
pH/ATC Combination Electrode | |
Mettler Toledo Five and Seven series pH meters | |
7 | |
0.01 | |
120 mm | |
12 mm |
BNC Waterproof, RCA | |
Epoxy Body | |
0°C to 100°C | |
ROSS electrode 3 M KCl fill solution (Cat. No. 810007) | |
ROSS | |
0.03 pH for buffers using automatic temperature compensation | |
2 Years | |
1 m |