Learn More
Thermo Scientific™ Orion™ Star Series RS232 Printer and Computer Cable Set
Connect your computer and printer to a Versa Star, Star A200 or Star A300 series meter using the Thermo Scientific™ Orion™ Star Series RS232 Printer and Computer Cable Set.
Brand:  Thermo Scientific™ 1010004
Product Code. 12043313
  • 76.78 € / Each
Estimated Shipment: 03-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Connect your computer and printer to a meter using the Thermo Scientific™ Orion™ Star Series RS232 Printer and Computer Cable Set. These cables are designed for use with Thermo Scientific™ Orion™ Versa Star meters, Thermo Scientific™ Orion™ Star A series meters (Star A320, Star A220 and Star A210 series) and legacy Orion™ Star and Star Plus meters.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Orion Star series RS232 printer and computer cable set
Meter Interface Cable
One RS232 printer interface cable (2.5mm RS232 meter connector and 25 pin RS232 printer connector) and one RS232 computer interface cable (2.5mm RS232 meter connector and 9 pin RS232 computer connector)
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.