missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Orion™ Silver/Sulfide Electrodes
Make quick, simple, accurate and economical measurement of silver and sulfide ions in aqueous solutions with these electrodes.
745.71 € - 888.42 €
Specifications
Drift | drift-free potentials |
---|---|
Junction | The free-flowing liquid junction assures stable |
Length (Metric) Cable | 1M |
For Use With (Equipment) | ISE Meters |
Temperature (Metric) Operating | 0°C to 80°C |
Product Code | Brand | Connector Types | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Connector Types | Price | Quantity & Availability | |||||
10664921
|
Thermo Scientific™
9416BN |
BNC |
745.71 €
Each |
Estimated Shipment: 12-12-2024
Log in to see stock. |
|||||
10374521
|
Thermo Scientific™
9616BNWP |
Waterproof BNC |
888.42 €
Each |
In Stock
Log in to see stock. |
|||||
Description
Make quick, simple, accurate and economical measurements of silver and sulfide ions in aqueous solutions with Thermo Scientific™ Orion™ Silver/Sulfide Electrodes. These electrodes have dual electrode-measuring silver and sulfide ions in aqueous solutions, and perform low-level cyanide and halide titrations.
The ThermoFisher Orion Silver/Sulfide electrode complies with SM 4500-S2- G, ASTM D4658-08 for Wastewater measurements.
Features
9616BNWP is a combination electrode with a Sure-Flow™ junction that allows easy cleaning. When conventional electrode junctions clog, the Sure-Flow junction can be opened and flushed clean simply by pressing the cap.
Silver/Sulfide Half-Cell Electrode 9416BN is to be used with the Orion 900200 sure flow double Junction reference electrode.
Solutions required
ISA
940011 ISA for silver measurements, 475 mL bottle
941609 SAOB II reagent pack for sulfide measurements, 4 x 475 mL bottle
Electrode fill solution
900002 Inner chamber filling solution for the double junction reference electrode,
5 x 60 mL bottles
900003 Outer chamber filling solution for the doublejunction reference electrode,
5 x 60 mL bottles
Titrant for sulphide titration
948206 0.1 M lead perchlorate solution, 475 mL bottles
Items included with
948201 Polishing strip. Qty 2.
Warranty period
12 months from date of purchase
Specifications
drift-free potentials | |
1M | |
0°C to 80°C |
The free-flowing liquid junction assures stable | |
ISE Meters |