missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Orion™ Sodium Electrodes

Measure sodium ions in aqueous solution quickly, accurately and economically.

Brand:  Thermo Scientific™ 8611BNWP


 View more versions of this product

Product Code. 10384141

  • 763.80 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Measure sodium in aqueous solutions quickly, accurately and economically with Thermo Scientific™ Orion™ Sodium Electrodes. A unique redox internal system with a Thermo Scientific™ Orion™ ROSS™ electrode provides faster response, and greater stability and accuracy than conventional sodium electrodes.

The sodium electrode is commonly used to measure samples such as food, beverages and animal feed.

Features
8611BNWP is a combination electrode with a Sure-Flow™ junction that allows easy cleaning. When conventional electrode junctions clog, the Sure-Flow junction can be opened and flushed clean simply by pressing the cap. 8411BN is a half cell sensor that requires ROSS reference electrode, Cat. No. 800300 or the ROSS Ultra reference electrode, Cat. No. 800500U to function.
 
Solutions required
Standards
941706     0.1 M NaCl sodium standard solution, 475 mL bottle
841108     1000 ppm sodium standard solution, 475 mL bottle
941107     100 ppm sodium standard solution, 475 mL bottle
941105     10 ppm sodium standard solution, 475 mL bottle
 
Known standards addition
841109     Sodium known addition standard, 1000 ppm with ISA, 475 mL bottle
650700     Sodium known addition standard, 1 M with ISA, 475 mL bottle
 
ISA
841111     Sodium ISA, 475 mL bottle
 
Electrode fill solution
900010    ROSS sodium and reference electrode filling solution, 5 x 50 mL bottles. 
                For regular sodium measurements (> 10-5 M or 0.2 ppm)
900012    ROSS sodium and reference electrode  filling solution, 5 x 50 mL bottles. 
                For low-level sodium measurements (< 10-5 M or 0.2 ppm)
 
841101     Sodium electrode storage solution, 475 mL bottle
 
Recondition solution 
841113     Sodium electrode reconditioning solution, 475 mL bottle
 
Items included with
841113     Sodium electrode reconditioning solution, 475 mL bottle
841111     Sodium ISA, 475 mL bottle
841109     Sodium known addition standard, 1000 ppm with ISA, 475 mL bottle
841108     1000 ppm sodium standard solution, 475 mL bottle
900010     ROSS sodium and reference electrode. 60mL 1 bottle
841101     Sodium electrode storage solution, 475 mL bottle
 
Warranty period
12 months from date of purchase by Buyer or 18 months from date of shipment from Seller, whichever is earlier.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Waterproof BNC
0.47 in
900010 for regular Na+ (> 10-5 M or 0.2 ppm)
900012 for low level Na+ (< 10-5 M or 0.2 ppm)
4.72 in
120 mm
Glass Body
32 to 176°F
0°C to 100°C
Sodium Combination Ion Selective Electrode
ROSS sodium combination electrode
12 mm
Sure flow
39.3 in.
1 M
Saturated to 10-6 M, saturated to 0.02ppm
ISE Meters
  • 1 x 10-6 M to saturated
  • 0.02 ppm to saturated
Yes
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • SODIUM SPECIFIC ELECTRODE

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 1
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 1 B
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H411-Toxic to aquatic life with long lasting effects.
  • H412-Harmful to aquatic life with long lasting effects.

Precautionary Statement
  • P202-Do not handle until all safety precautions have been read and understood.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P321-Specific treatment (see on this label).

Supplemental information
  • MIXTURE LIST-Contains : ammonium chloride, ammonium hydroxide, triton X-100

SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.