Learn More
Thermo Scientific™ Orion™ All-in-One™ pH Buffer Kits
Work with ease using this color-coded pH buffer kit that includes three buffers, storage solution and a storage bottle.
Brand: Thermo Scientific™ 910199
110.20 EUR valid until 2024-12-31
Use promo code "21745" to get your promotional price.
Description
Thermo Scientific™ Orion™ All-in-One™ pH buffer kits include pH 4.01, 7.00 and 10.01 buffers that are color-coded for easy identification as well as storage solution to help ensure your pH electrode is properly maintained. Using high-quality pH buffers to calibrate your pH electrode and pH meter is the best way to ensure your pH measurements are accurate and reproducible. Orion pH buffers are NIST traceable and manufactured under ISO 9001 quality standards.
Features:- pH buffers with temperature tables on the labels for accurate calibrations and NIST™ traceability
- Manufactured under ISO 9000 quality standards
- Color coded buffers: pH 4.01 buffer is pink; pH 7.00 buffer is yellow; and pH 10.01 buffer is blue
Includes: One 1 pint (475mL) bottle each of pH 4.00 buffer (910104), pH 7.00 buffer (910107), pH 10.01 buffer (910110) and standard pH electrode storage solution (910001) and one pH electrode storage bottle.
Specifications
Standard All-in-One pH Buffer Kit | |
4 x 475 mL | |
Kit | |
475 mL bottle each of pH 4.01, 7.00 and 10.01 buffers; 475 mL bottle of pH electrode storage solution (910001); and 12 mm diameter electrode storage bottle |
Accurate calibrations and NIST traceability, laboratory and bench analysis | |
pH Buffer and Storage Solution | |
Yes |
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.