Learn More
Thermo Scientific™ Orion Star™ A221 Portable pH Meter
BIOPHARMA
Reliably measure pH, mV, ORP and temperature with our portable pH meter for advanced field analysis and printer or computer interfacing.
Brand:  Thermo Scientific™ STARA2210
 View more versions of this product
Product Code. 11631469
  • 586.92 € / Each
Explore more special offers
Description
Description
Record accurate and reliable pH, mV, ORP and temperature measurements with the Thermo Scientific™ Orion Star™ A221 Portable pH Meter. This meter is ideal for a wide range of applications and advanced pH analysis in the field. Perform up to a 5 point pH calibration, log up to 1000 data point sets with time/date stamp and easily transfer calibration and data logs via USB or RS232 to a printer or computer with free downloadable Orion Star™ Com software. Waterproof meter ensures durable, rugged performance in outdoor environments with an IP67-rated housing.
Don’t leave your measurements to chance when analyzing samples outside the lab. Orion Star A221 Portable pH Meters are designed to offer the reliability you need in a user-friendly package for your dedicated pH testing needs. This waterproof, portable meter displays critical data with your measurement including temperature, date and time, read type, measurement stability, active calibration info and power source status.
  • Measure pH, mV, relative mV or ORP with temperature
  • Perform up to a 5 point pH calibration using automatic buffer recognition of NIST or DIN buffer groups with choice to manually enter custom buffer values
  • Informative, easy-to-read backlit graphic display with clear onscreen instructions simplifies operation
  • Capture your critical readings with measurement stability indicator and selectable read modes — Auto-Read holds a stable reading and Continuous shows changing readings with hold option
  • Data log collects and date/time stamps up to 1000 measurement sets
  • Non-volatile meter memory ensures your data and settings are protected, even with loss of power
  • Orion Star Com software facilitates data transfer to a computer via USB or RS232 connections
  • Flexible power options with AA batteries or optional universal power adapter
  • IP67-rated housing is waterproof for rugged field operation
  • Meter kits include hard-sided field case and protective meter armor with electrode holders, adjustable wrist/belt strap and built-in meter stand
  • Meters offer CE, TUV 3-in-1 and FCC Class A certifications
  • 3-year replacement warranty
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Universal 50-60 Hz, 100-240 VAC power adapter (sold separately, Cat. No. 1010003)
±0.2 mV or ±0.05% of reading whichever is greater
Calibration due alarm (fixed 12 hour)
800 hr.
pH, relative mV (RmV), ORP and temperature
CE, TUV 3-1, FCC Class A
1000 with date and time stamp
1
IP 67
±2000.0 mV
0.1/0.01/0.001
Star A221 pH meter, 4 AA batteries, computer cable
7.000pH
English, Spanish, French, Italian, German and Chinese; Portuguese and Korean available with software update
Non-volatile memory preserves data log, calibration log and meter settings
Orion Star A221 pH/mV/RmV/ORP/Temperature Portable Meter
pH electrode, ORP electrode, reference half-cell electrode, ATC temperature probe (purchase separately)
±2000.0 mV
0.1mV
±0.1°C
5°C to 45°C (ambient)
22.0°F to 221.0° F
Manual or automatic with ATC temperature probe
24cm x 10cm x 6.5cm (9.5 in. x 3.9 in. x 2.6 in.)
0.5 kg
USB, RS-232
±0.002
Yes, selectable
4 x AA
1 point relative mV (RmV) mode, 1 point ORP mode
Transfer single, range or all data points to printer or computer
Orion Star A221 pH portable meter
LCD Graphic, Backlit
pH, mV, Relative mV, ORP with Temperature
-2 to 20
-5.0°C to +105.0°C
BNC (pH or ORP electrode), pin-tip (reference electrode), 8 pin MiniDIN (ATC temperature probe)
Comprehensive with menu-specific function keys and dual purpose scroll/shortcut keys
Automatic data logging with Auto-Read measure mode; manual data logging with Continuous measure mode
10 per channel with password protection
4 AA batteries (included) or universal AC adapter (sold separately)
±2000.0 mV
5 to 85%, non-condensing
Smart Stability and Smart Averaging
41°F to 113°F (ambient)
1 point temperature offset calibration
0.1°C, 0.1°F
3 Year Meter
1.2 lb.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products