Learn More
Thermo Scientific™ Orion Star™ A321 pH Portable Meter
BIOPHARMA
Accurately monitor pH, mV, ORP and temperature with our waterproof, portable pH meter for high performance in challenging field locations.
864.30 € - 1256.25 €
Specifications
Outputs USB, RS-232
AC Adapter Universal 50-60 Hz, 100-240 VAC power adapter (sold separately, Cat. No. 1010003)
Accuracy (mV) ±0.2 mV or ±0.05% of reading whichever is greater
Accuracy (pH) ±0.002
Alarm Output High/low limit alarm, calibration due alarm
View More Specs
Products 2
Product Code Brand Description Includes Price Quantity & Availability  
Product Code Brand Description Includes Price Quantity & Availability  
11691469
BIOPHARMA
View accessory products View Documents
Thermo Scientific™
STARA3210
Orion Star A321 pH portable meter Star A321 pH meter, 4 AA batteries, computer cable
864.30 €
Each
11601479
BIOPHARMA
View accessory products View Documents
Thermo Scientific™
STARA3215
Orion Star A321 pH portable meter TRIS kit with ROSS Ultra low-maintenance pH/ATC electrode Star A321 pH meter, 8107UWMMD ROSS Ultra gel-filled epoxy-body pH/ATC electrode with 3m cable, pH 4/7/10 buffer and rinse pouches, ROSS storage solution, protective meter armor, hard-sided carrying case, 4 AA batteries, computer cable
1256.25 €
Each
Estimated Shipment: 03-02-2025
to see stock.
Add to basket
Description
Description
Get high accuracy and premium performance everywhere you need it with the Thermo Scientific™ Orion Star™ A321 pH Portable Meter. This meter is designed for a wide range of pH, mV, ORP and temperature testing and field applications. Evaluate samples in the most demanding locations using this waterproof meter with IP67-rated housing. Perform up to a 5 point pH calibration, log up to 5000 data point sets with time/date stamp and easily transfer calibration and data logs via USB or RS232 to a printer or computer with free downloadable Orion Star™ Com software.
Take your measurements at the source with our premium line of portable meters. The Thermo Scientific Orion Star A321 pH Portable Meter offers an information-rich screen and user-friendly operation. This rugged, waterproof portable meter is ready to tackle the elements with you. Orion Star A321 pH Portable Meters are designed to be easy-to-use, dependable field meters that offer advanced features and functions including a 5000 point date log with sample ID and user ID options, calibration and high/low limit alarms and password-protected measurement methods.
  • Measure pH, mV, relative mV or ORP with temperature
  • Perform up to a 5 point pH calibration using automatic buffer recognition of NIST or DIN buffer groups with choice to manually enter custom buffer values
  • Calibration editing option allows individual points to be fixed without a full recalibration
  • Informative, easy-to-read backlit graphic display with clear onscreen instructions simplifies operation
  • Capture your critical readings with measurement stability indicator and selectable read modes — Auto-Read holds a stable reading, Continuous shows changing readings with hold option and Timed logs data at specific intervals
  • Data log collects and date/time stamps up to 5000 measurement sets with optional sample ID and user ID
  • Calibration log stores 10 most recent calibrations
  • Non-volatile meter memory ensures your data and settings are protected, even with loss of power
  • Orion Star Com software facilitates data transfer to a computer via USB or RS232 connections
  • Flexible power options with AA batteries or optional universal power adapter
  • IP67-rated housing is waterproof for rugged field operation
  • Meter kits include hard-sided field case and protective meter armor with electrode holders, adjustable wrist/belt strap and built-in meter stand
  • Meters offer CE, TUV 3-in-1 and FCC Class A certifications
  • 3-year replacement warranty
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Specifications
USB, RS-232
±0.2 mV or ±0.05% of reading whichever is greater
High/low limit alarm, calibration due alarm
800 hr.
pH with calibration editing option, relative mV (RmV), ORP and temperature
CE, TUV 3-1, FCC Class A
5000 with date and time stamp
LCD Graphic, Backlit
pH, mV, Relative mV, ORP with Temperature
-2 to 20
-5.0°C to +105.0°C
7.000pH
English, Spanish, French, Italian, German and Chinese; Portuguese and Korean available with software update
Non-volatile memory preserves data log, calibration log and meter settings
Orion Star A321 pH/mV/RmV/ORP/Temperature Portable Meter
±2000.0 mV
5 to 85%, non-condensing
Selectable fast, medium, slow or Smart Stability setting; Selectable off or Automatic Smart averaging setting
41°F to 113°F (ambient)
1 point temperature offset calibration
0.1°C, 0.1°F
3 Year Meter
1.2 lb.
Universal 50-60 Hz, 100-240 VAC power adapter (sold separately, Cat. No. 1010003)
±0.002
Yes, selectable
4 x AA
1 to 5
Transfer single, range or all data points to printer or computer
1
IP 67
±2000.0 mV
0.1/0.01/0.001
BNC (pH or ORP electrode), pin-tip (reference electrode), 8 pin MiniDIN (ATC temperature probe)
Comprehensive with menu-specific function keys and dual purpose scroll/shortcut keys
Automatic data logging with Auto-Read and Timed measure modes; manual data logging with Continuous m
10 per channel with password protection
4 AA batteries (included) or universal AC adapter (sold separately)
±2000.0 mV
0.1mV
±0.1°C
5°C to 45°C (ambient)
22.0°F to 221.0° F
Manual or automatic with ATC temperature probe
24cm x 10cm x 6.5cm (9.5 in. x 3.9 in. x 2.6 in.)
0.5 kg