Learn More
Thermo Scientific™ Orion Star T900 Series Titrator Accessories
Minimize maintenance downtime with easy to install Orion Star T900 series titrator replacement parts. Orion Star T900 series titrator accessories expand functionality.
Brand:  Thermo Scientific™ START-TB3
 View more versions of this product
Product Code. 15882127
  • 33.37 € / Each
Estimated Shipment: 12-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Get accessories and replacement parts for the Orion Star T910 pH titrator, Orion Star T920 redox titrator, Orion Star T930 ion titrator and Orion Star T940 all-in-one titrator. These are designed to expand titrator functionality and minimize maintenance downtime.
•Select interchangable burette sizes of 10mL, 20mL or 50mL to customize your titrator
•Bottle holder helps secure titrant to prevent spills and offers flexible placement
•Color-coded tubing and connectors simplify tubing installation and maintenance
•Computer software is designed to make data transfer simple and reliable
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Orion Star T900 series drying tube for desiccant or carbon dioxide trap materials
Titrator accessory, reagent cap
Orion Star T910 pH titrator, Orion Star T920 redox titrator, Orion Star T930 ion titrator, Orion Star T940 all-in-one titrator