missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Orion Star T900 Series Titrator Accessories

Minimize maintenance downtime with easy to install Orion Star T900 series titrator replacement parts. Orion Star T900 series titrator accessories expand functionality.

Brand:  Thermo Scientific™ START-B20

 View more versions of this product

Product Code. 15892107

  • 431.15 € / Each
Estimated Shipment: 04-03-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Get accessories and replacement parts for the Orion Star T910 pH titrator, Orion Star T920 redox titrator, Orion Star T930 ion titrator and Orion Star T940 all-in-one titrator. These are designed to expand titrator functionality and minimize maintenance downtime.

•Select interchangable burette sizes of 10mL, 20mL or 50mL to customize your titrator
•Bottle holder helps secure titrant to prevent spills and offers flexible placement
•Color-coded tubing and connectors simplify tubing installation and maintenance
•Computer software is designed to make data transfer simple and reliable

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Orion Star T900 series 20ml burette
Titrator accessory, burette
Orion Star T910 pH titrator, Orion Star T920 redox titrator, Orion Star T930 ion titrator, Orion Star T940 all-in-one titrator
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.