Learn More
Thermo Scientific™ Orion™ 2 Cell Conductivity Probes
Perform applications in either the lab or the field with these versatile 2-electrode conductivity cells, available in a variety of materials and measurement ranges.
362.81 € - 809.03 €
Product Code | Brand | Measurement Ranges | Connector Types | For Use With (Equipment) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Measurement Ranges | Connector Types | For Use With (Equipment) | Price | Quantity & Availability | |||
11748196
|
Thermo Scientific™
011010 |
1 μS/cm to 200 mS/cm | 8-pin DIN | 150Aplus, 145Aplus, 125Aplus, 115Aplus, 105Aplus Meters | N/A | ||||
10153012
|
Thermo Scientific™
011010A |
1 μS/cm to 200 mS/cm | 8-pin Waterproof DIN | 555A, 550A, 162A, 136A, 135A, 131S, 130A Meters |
615.06 €
Each |
Estimated Shipment: 04-02-2025
Log in to see stock. |
|||
11768196
|
Thermo Scientific™
011020 |
0.1 μS/cm to 100 mS/cm | 8-pin DIN | 150Aplus, 145Aplus, 125Aplus, 115Aplus, 105Aplus Meters |
585.92 €
Each |
Estimated Shipment: 07-02-2025
Log in to see stock. |
|||
10727564
|
Thermo Scientific™
011020A |
0.1 to 100 μS/cm | 8-pin DIN | 555A, 550A, 162A, 136A, 135A, 131S, 130A Meters |
809.03 €
Each |
Estimated Shipment: 04-02-2025
Log in to see stock. |
|||
10256152
|
Thermo Scientific™
011050 |
1 μS/cm to 20 mS/cm | 8-pin DIN | 150Aplus, 145Aplus, 125Aplus, 115Aplus, 105Aplus Meters |
478.38 €
Each |
Estimated Shipment: 07-02-2025
Log in to see stock. |
|||
15937431
|
Thermo Scientific™
011050 |
1 μS/cm to 20 mS/cm | 8-pin DIN | 150Aplus, 145Aplus, 125Aplus, 115Aplus, 105Aplus Meters | N/A | ||||
10511833
|
Thermo Scientific™
011050MD |
1 μS/cm to 20 mS/cm | Mini-DIN | Orion Star Series Meters |
456.27 €
Each |
In Stock
Log in to see stock. |
|||
10770085
|
Thermo Scientific™
011510MD |
10 μS/cm to 200 mS/cm | 8-pin Mini-DIN | Orion Star Series Meters |
362.81 €
Each |
Estimated Shipment: 04-02-2025
Log in to see stock. |
|||
11996079
|
Thermo Scientific™
013016MD |
0.01 to 300 μS/cm | Mini-DIN | Orion Star Series Meters |
774.86 €
Each |
In Stock
Log in to see stock. |
|||
10113042
|
Thermo Scientific™
018020MD |
10 μS/cm to 2000 mS/cm | Mini-DIN | Orion Star Series Meters |
567.83 €
Each |
In Stock
Log in to see stock. |
|||
Description
Achieve accurate measurements from ultrapure water, sea water, and more with Thermo Scientific™ Orion™ 2-Electrode Cells. These probes use two electrode cells to measure low-, standard-, and high-conductivity samples based on the cell constant. They are a versatile option for precise measurements in the lab and field. 013016MD is ideal in low ionic strength solutions, such as deionized water and ultrapure water when paired with a flow-cell assembly. 018020MD is good for chemically reactive conductivity samples, including concentrated acids and bases. 011050MD and 011510MD epoxy/graphite cells are durable, chemical resistant, and measure a wide range of conductivities in the lab or field.
The ThermoFisher Orion Conductivity electrode 013016MD complies with SM 2510B; D1125-95, -99 (A) for Drinking water measurements and USP<645> Measuring the Conductivity of Pure WaterFeatures
These probes use two electrode cells to measure low-, standard-, and high-conductivity samples based on the cell constant. They are a versatile option for precise measurements in the lab and field.
013016MD is ideal in low ionic strength solutions, such as deionized water and ultrapure water when paired with a flow-cell assembly . 018020MD is good for chemically reactive conductivity samples, including concentrated acids and bases. 011050MD and 011510MD epoxy/graphite cells are durable, chemical resistant, and measure a wide range of conductivities in the lab or field.
Solutions required
Standards
011008 100 µS/cm conductivity/TDS standard, 5 x 60 mL bottles
01100910 147 µS/cm conductivity standard, 10 pouches
011007 1413 µS/cm conductivity/TDS standard, 5 x 60 mL bottles
01100710 1413 µS/cm conductivity/TDS standard, 10 pouches
011006 12.9 mS/cm conductivity/TDS standard, 5 x 60 mL bottles
01100610 12.9 mS/cm conductivity/TDS standard, 10 pouches
011005 111 mS/cm conductivity standard, 5 x 60 mL bottles
01100510 111 mS/cm conductivity standard, 10 pouches
Conditioning solution
011001 Conditioning solution for 011050MD conductivity cells
Accessories
013017 Replacement flow cell
Items included
011050MD 011007 1413 µS/cm conductivity/TDS standard, 60 mL 1 bottle
011001 Conditioning solution, 60 mL 1 bottle
011510MD 011006 12.9 mS/cm conductivity/TDS standard, 60 mL 1 bottle
011007 1413 µS/cm conductivity/TDS standard, 60 mL 1 bottle
013016MD 011008 100 µS/cm conductivity/TDS standard, 60 mL 1 bottle
013017 Replacement flow cell
Warranty period
12 months from date of purchase
Specifications
2-Electrode Conductivity Cells |