missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Orion™ Versa Star Pro™ pH/Conductivity Multiparameter Benchtop Meter
BIOPHARMA

Get premium performance and flexibility with our benchtop meter with pH and conductivity modules for simultaneous pH, conductivity, and temperature.

Brand:  Thermo Scientific™ VSTAR50

1997.50 EUR valid until 2024-12-15
Use promo code "21658" to get your promotional price.


 View more versions of this product

Product Code. 11611509

  • 2422.05 € / Each
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Get high accuracy, premium performance and ultimate flexibility with the Thermo Scientific™ Orion™ Versa Star Pro™ pH/Conductivity Multiparameter Benchtop Meter. Meet your most challenging applications for pH, mV, ORP, conductivity, total dissolved solids (TDS), salinity, resistivity and temperature. Easily view measurements on the stunning color display and customize the four meter channels with interchangeable modules. View pH and conductivity simultaneously or view each channel separately. Log up to 2000 data point sets with time/date stamp and easily transfer logs to a printer or computer.

Thermo Scientific Orion Versa Star Pro Benchtop Meters are designed to meet your most challenging application with ease. Delivering rich functionality and features, you can optimally configure your meter system to meet changing requirements. The Orion Versa Star Pro Benchtop Meters offer interchangeable measurement modules that allow multiple users to customize four separate channels to meet their specific requirements. Easily view your measurements on a stunning color display with adjustable text sizes. Take the guesswork out of meter calibration and setup with onscreen instructions available in a variety of local languages. Enjoy quick and simple data entry using the numeric keypad.
  • Onscreen measurement stability indicator lets you know exactly when your measurements are ready
  • Selectable read modes include Auto-Read to hold a stable reading, Continuous to show changing readings, Timed to log data at specific intervals and Single-Shot to hold a reading after a specific wait time is reached
  • Audible alarms automatically notify you when a set offset value, high/low limit value, and calibration time interval are reached
  • Easily access, update and switch your preferred measurement settings, using 10 password-protected methods per channel
  • Uniformly mix solutions using up to two meter-controlled stirrer probes that are easy to position and can be quickly rinsed between samples
  • Data log collects and date/time stamps up to 2000 measurement sets with optional electrode ID, sample ID and account ID
  • Calibration log stores 30 most recent calibrations per parameter with viewable graph for multipoint pH and ISE calibrations
  • Ensure measurement integrity by viewing the active calibration log while taking readings
  • Non-volatile meter memory preserves data and settings, even in the event of power loss
  • Meter facilitates data transfers to a computer via USB or RS232 connections
  • IP54-rated housing resists dust and splashes and can be wall-mounted if table space is limited
  • Meters include CE, TUV 3-in-1 and FCC Class A certifications
  • 3-year replacement warranty
Includes Orion Versa Star pH Measurement Module:
  • Measure pH, mV, relative mV or ORP with temperature
  • Perform up to a 6 point pH calibration using automatic buffer recognition of NIST, DIN or user-defined buffer groups with choice to manually enter custom buffer values
  • Calibration editing option allows individual points to be fixed without a full recalibration
Includes Orion Versa Star Conductivity Measurement Module:
  • Measure conductivity, TDS, salinity or resistivity with temperature
  • Perform up to a 6 point conductivity calibration with calibration editing option that allows individual points to be fixed without a full recalibration
  • Auto-ranging conductivity values offer consistent readings across entire measurement range without need to recalibrate
  • Use linear, non-linear, EP (USP) or off conductivity temperature compensation with 5°C, 10°C, 15°C, 20°C or 25°C reference temperatures

System Access Feature
Orion Versa Star Pro meters offer a system access feature that is designed to help ensure data integrity by preventing undesired access to various meter functions including date/time setting, data log and calibration log. The System Access feature can be easily turned on or off as the needs of your lab change for ultimate flexibility in operation and ease-of-use. When enabled, the System Access feature allows assignment of a system administrator and up to 10 user accounts with customizable access to instrument settings, measurement settings, calibrations and logs. Use the password protection to secure selected meter functions, while leaving other functions accessible for all users. The System Access feature allows you to customize meter access to meet your specific security and data reporting needs.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Universal 50-60 Hz, 100-240 VAC power adapter (Cat. No. STARA-PWR)
0.5% of reading ±1 digit > 3 μS; 0.5% of reading ±0.01 μS ≤ 3 μS
±0.002
0.5% of reading ±1 digit
Set point alarm, high/low limit alarm, calibration due alarm
pH with calibration editing option, relative mV (RmV), ORP, conductivity with calibration editing op
Up to 6
0.001 to 199.9cm-1
Transfer single, range or all data points to printer or computer
Orion Versa Star Pro benchtop meter with pH/mV module, conductivity/TDS/salinity/resistivity module and stand
Color graphic LCD with user-customizable options
pH, mV, Relative mV, TDS, ORP with Temperature, Salinity, Conductivity, Resistivity with Temperature
±2000.0 mV
2 ohm to 100.0 meg-ohm
0.001 ppm to 200.0 ppt
-5.0°C to +105.0°C
BNC (pH or ORP electrode), pin-tip (reference electrode), 8 pin MiniDIN (ATC temperature probe), 8 pin MiniDIN (conductivity sensor with built-in temperature), two stirrer inputs (stirrer probe)
Numeric with menu-specific function keys and dual purpose scroll/shortcut keys
Automatic data logging with Auto-Read, Timed and Single Shot measure modes; manual data logging with
10 per channel
Universal AC adapter (included)
±2000.0 mV
0.001 μS minimum, auto ranging up to 4 significant digits
1 ohm or 0.1 meg-ohm, auto ranging
0.001 ppm minimum, auto ranging up to 4 significant digits
Selectable fast, medium, slow or Smart Stability setting; Selectable off or Automatic Smart averaging setting
±0.1°C
41°F to 113°F (ambient)
5, 10, 15, 20, 25°C
22.0°F to 221.0° F
Manual or automatic with ATC temperature probe
28cm x 24cm x 10cm (11 in. x 9.5 in. x 3.9 in.), modules add ≤ 2cm (0.8 in.) L & H
1.2 kg, modules add 0.2 kg each
USB (2), RS232
±0.2 mV or ±0.05% of reading whichever is greater
0.5% of reading ±1 digit
0.5% of reading ±1 digit
Yes
1 point relative mV (RmV) mode, 1 point ORP mode
1 to 6
CE, TUV 3-1, FCC Class A
2000 with date and time stamp
4 - customizable with modules
IP 54
0.001 μS/cm to 3000 mS/cm
-2.000 to 20.000
0.06 to 80.00 psu, 0.05 to 42.00 ppt
0.1/0.01/0.001
Versa Star Pro meter, VSTAR-PH pH module, VSTAR-CND conductivity module, electrode stand, universal power adapter, literature CD, printed quick start guide, computer interface cable and meter test certificate
7.000pH (adjustable)
English, Spanish, French, Italian, German, Chinese, Portuguese and Korean
Non-volatile memory preserves data log, calibration log and meter settings
DC input: 9VDC, 1A
±2000.0 mV
5 to 85 %, non-condensing
0.1mV
0.01 psu or 0.01 ppt minimum, auto ranging
Practical salinity (psu) or natural sea water (ppt)
Linear (0.02 to 9.99)
Linear, nLFn (non-linear ultra pure water), nLFu (non-linear ultra pure degassed water), EP (off with warning) or off
5°C to 45°C (ambient)
1 point temperature offset calibration
0.1°C, 0.1°F
3 year meter and module warranty
2.5 lb., modules add 0.5 lb. each
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers