missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ ORP Electrodes

Robust and reliable maintenance free solid electrolyte pH electrode for measurement up to 110 ° C that is ideal for industrial applications.

Brand:  Fisherbrand™ BP935-DSP-B120

Product Code. 11745618

  • 177.00 € / Each
Estimated Shipment: 10-01-2025
to see stock.
Explore more special offers
Add to basket

Description

Description

  • Maintenance free solid electrolyte pH electrode for measurement up to 110 ° C and 10 bar
  • Autoclavable 121 ° C
  • Length 12 or 22.5 cm
  • External thread PG 13,5 for in-line mounting
  • Ideal for Industrial applications
Specifications

Specifications

ORP Electrode
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers