missing translation for 'onlineSavingsMsg'
Learn More

Orthophosphoric Acid, 85+%, Extra Pure, SLR, Fisher Chemical™

31.70 € - 585.00 €

Chemical Identifiers

CAS 7664-38-2
Molecular Formula H3O4P
Molecular Weight (g/mol) 97.994
MDL Number 11340
InChI Key NBIIXXVUZAFLBC-UHFFFAOYSA-N
Synonym orthophosphoric acid, sonac, o-phosphoric acid, phosphorsaeure, acidum phosphoricum, evits, wc-reiniger, acide phosphorique, white, acido fosforico
PubChem CID 1004
ChEBI CHEBI:26078
IUPAC Name phosphoric acid
SMILES OP(=O)(O)O
View More Specs

Products 5
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10122010
SDS View Documents
Fisher Chemical
O/0450/PB08
500 mL HDPE plastic bottle
31.70 €
500mL
Please to purchase this item. Need a web account? Register with us today!
10172442
SDS View Documents
Fisher Chemical
O/0450/PB15
1 L HDPE plastic bottle
56.55 €
1 L
Please to purchase this item. Need a web account? Register with us today!
10182050
SDS View Documents
Fisher Chemical
O/0450/PB17
2.5 L HDPE plastic bottle
91.20 €
2.50 L
Please to purchase this item. Need a web account? Register with us today!
10020662
SDS View Documents
Fisher Chemical
O/0450/PB21
5 L HDPE plastic bottle
121.00 €
5 L
Please to purchase this item. Need a web account? Register with us today!
10294052
SDS View Documents
Fisher Chemical
O/0450/25
25 L Plastic container
585.00 €
25 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

7664-38-2
97.994
NBIIXXVUZAFLBC-UHFFFAOYSA-N
1004
phosphoric acid
H3O4P
11340
orthophosphoric acid, sonac, o-phosphoric acid, phosphorsaeure, acidum phosphoricum, evits, wc-reiniger, acide phosphorique, white, acido fosforico
CHEBI:26078
OP(=O)(O)O

Specifications

158°C
7664-38-2
Liquid
11340
NBIIXXVUZAFLBC-UHFFFAOYSA-N
phosphoric acid
1004
2 hPa at 20°C
21°C
2
H3O4P
orthophosphoric acid, sonac, o-phosphoric acid, phosphorsaeure, acidum phosphoricum, evits, wc-reiniger, acide phosphorique, white, acido fosforico
OP(=O)(O)O
97.994
CHEBI:26078
Vapor Density: 3.4
Videos
Safety and Handling

Safety and Handling

Signal Word
  • Danger

Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1

Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

Supplemental information
  • MIXTURE LIST-Contains : Orthophosphoric acid

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.