missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OrthoPhosphoric Acid Solution 1M, Certified, for HPLC, Fisher Chemical
Shop All Fisher Chemical Products
Click to view available options
Quantity:
500 mL
Packaging:
Plastic Coated Glass bottle
Unit Size:
500mL
Specifications
Specifications
| CAS | 7664-38-2 |
| Physical Form | Liquid |
| Quantity | 500 mL |
| Molecular Formula | H3O4P |
| MDL Number | 11340 |
| Synonym | orthophosphoric acid, sonac, o-phosphoric acid, phosphorsaeure, acidum phosphoricum, evits, wc-reiniger, acide phosphorique, white, acido fosforico |
| InChI Key | NBIIXXVUZAFLBC-UHFFFAOYSA-N |
| SMILES | OP(=O)(O)O |
| IUPAC Name | phosphoric acid |
| Molecular Weight (g/mol) | 97.994 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction