missing translation for 'onlineSavingsMsg'
Learn More

Osmium, 99.9%, (trace metal basis)

Product Code. 10045483 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 10045483

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7440-04-2 | Os | 190.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for ultra trace analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4-2-7440
Molecular Formula Os
Molecular Weight (g/mol) 190.23
MDL Number MFCD00011147
InChI Key SYQBFIAQOQZEGI-UHFFFAOYSA-N
Synonym ion, metallic, osmium, elemental, unii-2e7m255opy, os4, osmio, hydrido, hydride, atom, powder
PubChem CID 23937
ChEBI CHEBI:30687
IUPAC Name osmium
SMILES [Os]

Specifications

Color Black
Melting Point 3045.0°C
Boiling Point 5027.0°C
Quantity 5 g
Packaging Glass bottle
Assay Percent Range 99.90%
Form Powder
Linear Formula Os
Merck Index 15, 6988
Solubility Information Solubility in water: insoluble.
Formula Weight 190.2
Percent Purity 99.9%
Grade Trace Metal Basis
Chemical Name or Material Osmium
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.