missing translation for 'onlineSavingsMsg'
Learn More

Osmium powder, -200 mesh, 99.8% (metals basis), Thermo Scientific Chemicals

154.00 € - 562.00 €

Chemical Identifiers

CAS 7440-04-2
Molecular Formula Os
Molecular Weight (g/mol) 190.23
MDL Number MFCD00011147
InChI Key SYQBFIAQOQZEGI-UHFFFAOYSA-N
Synonym ion, metallic, osmium, elemental, unii-2e7m255opy, os4, osmio, hydrido, hydride, atom, powder
PubChem CID 23937
ChEBI CHEBI:30687
IUPAC Name osmium
SMILES [Os]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11301809
View Documents Promotion Details
Thermo Scientific Alfa Aesar
012446.03
1 g
154.00 €
1g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
11391799
View Documents Promotion Details
Thermo Scientific Alfa Aesar
012446.06
5 g
562.00 €
5g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7440-04-2
190.23
SYQBFIAQOQZEGI-UHFFFAOYSA-N
23937
osmium
Os
MFCD00011147
ion, metallic, osmium, elemental, unii-2e7m255opy, os4, osmio, hydrido, hydride, atom, powder
CHEBI:30687
[Os]

Specifications

7440-04-2
MFCD00011147
ion, metallic, osmium, elemental, unii-2e7m255opy, os4, osmio, hydrido, hydride, atom, powder
[Os]
190.23
CHEBI:30687
−200 Mesh
Powder
Osmium powder
Os
UN3089
SYQBFIAQOQZEGI-UHFFFAOYSA-N
osmium
23937
99.8%
Strong
(metals basis)
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H228
Flammable solid.

P210-P240-P241-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P310-P312-P332+P313-P362-P370+P378q-P501c

H228-H315-H318-H335

missing translation for 'dotInformation' : Transport Hazard Class: 4.1; Packing Group: II; Proper Shipping Name: METAL POWDERS, FLAMMABLE, N.O.S.

missing translation for 'einecsNumber' : 231-114-0

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Certificates

RUO – Research Use Only