Learn More
Osmium tetroxide, 99.9+%, (trace metal basis), Thermo Scientific Chemicals
52.06 € - 1107.51 €
Chemical Identifiers
CAS | 20816-12-0 |
---|---|
Molecular Formula | O4Os |
Molecular Weight (g/mol) | 254.23 |
MDL Number | MFCD00011150 |
InChI Key | VUVGYHUDAICLFK-UHFFFAOYSA-N |
Synonym | osmium tetroxide, osmium tetraoxide, osmic acid, perosmic oxide, osmic acid anhydride, osmium viii oxide, oso4, perosmic acid anhydride, osmiumtetroxide, osmium oxide, t-4 |
PubChem CID | 30318 |
IUPAC Name | tetraoxoosmium |
SMILES | O=[Os](=O)(=O)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10042610
|
Thermo Scientific Acros
191181000 |
100 mg |
52.06 €
100mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10531641
|
Thermo Scientific Acros
191182500 |
250 mg |
96.78 €
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10760701
|
Thermo Scientific Acros
191185000 |
500 mg |
167.84 €
500mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10174350
|
Thermo Scientific Acros
191180010 |
1 g |
283.41 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10207243
|
Thermo Scientific Acros
191180050 |
5 g |
1107.51 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysis, Oxidation reagentChemical Identifiers
20816-12-0 | |
254.23 | |
VUVGYHUDAICLFK-UHFFFAOYSA-N | |
30318 | |
O=[Os](=O)(=O)=O |
O4Os | |
MFCD00011150 | |
osmium tetroxide, osmium tetraoxide, osmic acid, perosmic oxide, osmic acid anhydride, osmium viii oxide, oso4, perosmic acid anhydride, osmiumtetroxide, osmium oxide, t-4 | |
tetraoxoosmium |
Specifications
130.0°C | |
20816-12-0 | |
Crystals | |
O4Os | |
MFCD00011150 | |
15, 6990 | |
Passes test | |
O=[Os](=O)(=O)=O | |
254.23 | |
254.2 | |
Trace Metal Basis | |
Osmium tetroxide |
40.0°C to 42.0°C | |
Colorless to Yellow | |
99.9+% | |
OsO4 | |
01,759; 02,301; 04,361; 05,141; 06,424; 10,290; 12,358; 13,186; 14,235; 15,240; 16,249; 17,236 | |
osmium tetroxide, osmium tetraoxide, osmic acid, perosmic oxide, osmic acid anhydride, osmium viii oxide, oso4, perosmic acid anhydride, osmiumtetroxide, osmium oxide, t-4 | |
VUVGYHUDAICLFK-UHFFFAOYSA-N | |
tetraoxoosmium | |
30318 | |
≥99.9% | |
Ampoule |
Safety and Handling
GHS H Statement
Fatal in contact with skin.
Causes severe skin burns and eye damage.
Fatal if inhaled.
Fatal if swallowed.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
GHS P Statement
Rinse mouth.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/f
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 244-058-7
- Osmium(VIII)-tetroxide
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 2
- Acute toxicity Category 1
- Skin corrosion/irritation Category 1 B
- Serious eye damage/eye irritation Category 1
- Respiratory sensitiser Category 1
Hazard Statement
- H300-Fatal if swallowed.
- H310-Fatal in contact with skin.
- H330-Fatal if inhaled.
- H314-Causes severe skin burns and eye damage.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
- P330-Rinse mouth.
- P310-Immediately call a POISON CENTER/doctor/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.