missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Osmium Tetroxide, ACS Reagent, ≥98%, Honeywell Fluka™
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Glass Bottle
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
Boiling Point | 130°C |
Melting Point | 39.5°C to 41°C |
Physical Form | Solid |
Quantity | 250 mg |
Linear Formula | OsO4 |
UN Number | UN2471 |
Solubility Information | Slightly soluble in water |
Formula Weight | 254.23g/mol |
Percent Purity | ≥98% |
Odor | Unpleasant |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction