missing translation for 'onlineSavingsMsg'
Learn More

Osmium Tetroxide Solution, Honeywell Fluka™

Product Code. 15648330
Click to view available options
Quantity:
5 mL
25 mL
Packaging:
Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

2.5 wt. % in tert-butanol

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20816-12-0
Molecular Formula O4Os
Molecular Weight (g/mol) 254.23
MDL Number MFCD00011150
InChI Key VUVGYHUDAICLFK-UHFFFAOYSA-N
Synonym osmium tetroxide, osmium tetraoxide, osmic acid, perosmic oxide, osmic acid anhydride, osmium viii oxide, oso4, perosmic acid anhydride, osmiumtetroxide, osmium oxide, t-4
PubChem CID 30318
IUPAC Name tetraoxoosmium
SMILES O=[Os](=O)(=O)=O

Specifications

Physical Form Liquid
Quantity 5 mL
Linear Formula OsO4
UN Number UN3286
Flash Point 4°C (39.2°F)
Formula Weight 254.23g/mol
Packaging Glass Bottle
Vapor Pressure 6.76psi (55°C), 5.42psi (20°C)
Density 1.0g/mL at 20°C, 0.811g/mL at 25°C
Chemical Name or Material Osmium Tetroxide Solution, Contains tert-butyl hydroperoxide as stabilizer
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.