missing translation for 'onlineSavingsMsg'
Learn More

Osmium(VIII) oxide, 4% aq. soln., Thermo Scientific Chemicals

Product Code. 15472237
Click to view available options
Quantity:
2 mL
10 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

Osmium(VIII) oxide is widely used as an oxidant in organic synthesis to oxidize alkenes to vicinal diols as well as in the sharpless oxyamination to give vicinal amino-alcohols. It is involved in the oxidative cleavage of alkenes (Lemieux-Johnson oxidation) in association with sodium periodate. It finds application in transmission electron microscopy (TEM) as a staining agent. It is involved in the promotion of oxidative cleavage of graphene layers in carbon nanotubes and as an intermediate in osmium ore refining. As a lipid stain, it is useful in scanning electron microscopy (SEM) as an alternative to sputter coating.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20816-12-0
Molecular Formula O4Os
Molecular Weight (g/mol) 254.23
MDL Number MFCD00011150
InChI Key VUVGYHUDAICLFK-UHFFFAOYSA-N
Synonym osmium tetroxide, osmium tetraoxide, osmic acid, perosmic oxide, osmic acid anhydride, osmium viii oxide, oso4, perosmic acid anhydride, osmiumtetroxide, osmium oxide, t-4
PubChem CID 30318
IUPAC Name tetraoxoosmium
SMILES O=[Os](=O)(=O)=O

Specifications

Physical Form Liquid
Quantity 10 mL
Solubility Information Soluble in alcohol,ether,chloroform,benzene,ammonium hydroxide,phosphorus oxychloride and carbon tetrachloride
Formula Weight 254.1
Sensitivity Air Sensitive
Vapor Pressure 17mm Hg at 20°C
Chemical Name or Material Osmium(VIII) oxide
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Osmium(VIII) oxide, 4% aqueous solution
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H310-Fatal in contact with skin.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H332-Harmful if inhaled.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Osmium(VIII) oxide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.