missing translation for 'onlineSavingsMsg'
Learn More

Osmium(VIII) oxide, 99.8% (metals basis), Os 74.4% min, Thermo Scientific Chemicals

 

136.68 € - 1809.00 €

Chemical Identifiers

CAS 20816-12-0
Molecular Formula O4Os
Molecular Weight (g/mol) 254.23
MDL Number MFCD00011150
InChI Key VUVGYHUDAICLFK-UHFFFAOYSA-N
Synonym osmium tetroxide, osmium tetraoxide, osmic acid, perosmic oxide, osmic acid anhydride, osmium viii oxide, oso4, perosmic acid anhydride, osmiumtetroxide, osmium oxide, t-4
PubChem CID 30318
IUPAC Name tetraoxoosmium
SMILES O=[Os](=O)(=O)=O
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11346439
SDS View Documents
Thermo Scientific Alfa Aesar
012103.02
0.5 g
136.68 €
0.50g
Please to purchase this item. Need a web account? Register with us today!
11356439
SDS View Documents
Thermo Scientific Alfa Aesar
012103.03
1 g
225.12 €
1g
Please to purchase this item. Need a web account? Register with us today!
11366439
SDS View Documents
Thermo Scientific Alfa Aesar
012103.48
10 x 1 g
1809.00 €
10g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Oxidation, catalyst in Sharpless dihydroxylations Osmium(VIII) oxide is used in the Sharpless oxyamination to give vicinal amino-alcohols. It is used to oxidize alkenes to the vicinal diols. It acts as a staining agent used in transmission electron microscopy to provide contrast to the image and as a stain for lipids in optical microscopy. As a polymer stain, it is also used to stain selectively copolymers especially styrene-butadiene block copolymers. Further, it is involved in the synthesis of Isosteviol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

 
Specifications

Chemical Identifiers

20816-12-0
254.23
VUVGYHUDAICLFK-UHFFFAOYSA-N
30318
O=[Os](=O)(=O)=O
O4Os
MFCD00011150
osmium tetroxide, osmium tetraoxide, osmic acid, perosmic oxide, osmic acid anhydride, osmium viii oxide, oso4, perosmic acid anhydride, osmiumtetroxide, osmium oxide, t-4
tetraoxoosmium

Specifications

130°C
20816-12-0
99.8% (metals basis)
MFCD00011150
Soluble in chloroform,alcohol and ethers.Soluble in water,organic solvents,benzene,alcohol,ether,ammonium hydroxide,phosphorus oxychloride and carbon tetrachloride.
O=[Os](=O)(=O)=O
254.23
254.1
Crystalline
Osmium(VIII) oxide
40.6°C
Crystalline
O4Os
osmium tetroxide, osmium tetraoxide, osmic acid, perosmic oxide, osmic acid anhydride, osmium viii oxide, oso4, perosmic acid anhydride, osmiumtetroxide, osmium oxide, t-4
VUVGYHUDAICLFK-UHFFFAOYSA-N
tetraoxoosmium
30318
Like chlorine
10 hPa (8mm Hg) at 20°C
Videos
Safety and Handling

Safety and Handling

Keep container tightly closed and in a well-ventilated place. In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible).

P260-P262-P264b-P270-P271-P280-P284-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c

H300+H310+H330-H314-H335

missing translation for 'dotInformation' : DOT Class: 6.1, Packaging Group: I

missing translation for 'einecsNumber' : 244-058-7

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.