Learn More
Osmolality Standard 3000 mOsm/Kg, Reagecon
Used to calibrate, control or validate all types of osmometers.
Brand: Reagecon RE-OSM-3000
This item is not returnable.
View return policy
Description
- Osmolality Reference Standard, Osmolality Reference Material.
- The Osmolality results are reported in milliOsmoles per kilogram water (mOsm/kg H2O shortened to mOsm/kg for routine use).
- This product is manufactured in compliance with European, United States and Chinese Pharmacopoeia guidelines where appropriate.
- Certified by Reagecon’s ISO 17025 Accredited Test Method (INAB Ref 264T).
- Prepared gravimetrically on a weight/weight basis.
- Both solute and solvent are weighed on a balance calibrated by Reagecon engineers using OIML traceable weights.
- Reagecon holds ISO 17025 accreditation for calibration of laboratory balances (INAB Ref:265C).
- The resulting Balance Certificate of Calibration is issued in accordance with the requirements of ISO/IEC 17025.
- The certified values of each standard are verified using a high-performance osmometer calibrated with high quality reference materials.
Specifications
3000 mOsm/Kg | |
12 x 5 mL | |
Liquid |
Amber Glass Ampule | |
Calibration of Osmometer |