missing translation for 'onlineSavingsMsg'
Learn More

Buerkle™ OTAL™ Foot Pump made of PVDF

Ensure removal of residual liquid with this foot pump. OTAL™ Foot Pumps feature delivery tubes made of PVDF, ideal for acids and alkalis. Telescopic immersion tube adapts itself to the container height on insertion of the foot pump.

Brand:  Buerkle™ 5000-4016

Product Code. 10027580

  • 214.00 € / Each
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Simple handling: Both hands remain free for filling
  • Foot pump can be used with different containers
  • Very good pump capacity due to large volume foot bellows
  • With universal bungs made of soft PVC, for container openings of 30 to 70mm dia.
  • With its special material properties, PVDF has an even better chemical resistance than polypropylene
  • Resistant to chlorine bleach
  • Not for heavily oxidizing acids and alkalis
Specifications

Specifications

Industrial Sample Handling of Acids and Alkalis, Not for Heavily Oxidising Acids and Alkalis
Chlorine Bleach
Universal bungs made of PVC, for container openings of 30 mm to 70 mm dia.
PVDF
20 L/min.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers