Learn More
Hughes Outdoor Self-Draining Safety Shower with Eye/Face Wash
This ANSI compliant self-draining emergency safety shower with open stainless steel eye/face wash, suitable for hot climates.
1195.00 € - 1710.00 €
Specifications
Activation Push Lever, Foot Treadle
Product Type Safety Shower
View More Specs
Products 2
Product Code Brand Material Price Quantity & Availability  
Product Code Brand Material Price Quantity & Availability  
17722348
View Documents
Hughes
HSHDSD18GV
Galvanized Steel
1195.00 €
Each
Estimated Shipment: 01-05-2025
to see stock.
Add to basket
17732348
View Documents
Hughes
HSHDSD18SS
Stainless Steel
1710.00 €
Each
Estimated Shipment: 01-05-2025
to see stock.
Add to basket
Description
Description
  • This ANSI compliant self-draining emergency safety shower with open stainless steel eye/face wash, suitable for hot climates
  • The design allows standing water to drain away from the standpipe to ensure the delivery of tepid water
  • Fitted with foot panel as standard
  • The valve is situated directly above the water inlet; the bleed pipe is fitted above this so standing water can drain into the ground preventing stagnant water overheating from solar radiation and removing the risk of further injury to the operator
  • Available with galvanised powder coated mild steel (G) or stainless steel (GS) pipework
Specifications
Specifications
Push Lever, Foot Treadle
Safety Shower
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Hughes Outdoor Self-Draining Safety Shower with Eye/Face Wash