Learn More
Cytiva Outlet Valve
Brand:  Cytiva 29023898
Product Code. 15219774
  • 387.00 € / Each
Explore more special offers
Description
Description
For use with the AKTA start automated protein purification system. The outlet valve is used to direct the flow to the fraction collector or to the waste.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Direct the flow to the fraction collector or to the waste
AKTA start Automated Protein Purification System