missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ OVA257-264 (SIINFEKL) peptide bound to H-2Kb Monoclonal Antibody (eBio25-D1.16 (25-D1.16)), Brilliant Violet™ 650, eBioscience™, Invitrogen™

Mouse Monoclonal Antibody

188.00 € - 385.00 €

Specifications

Antigen OVA257-264 (SIINFEKL) peptide bound to H-2Kb
Clone eBio25-D1.16 (25-D1.16)
Concentration 0.2 mg/mL
Applications Flow Cytometry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17932031
View Documents
Invitrogen™
416-5743-82
100 μg
385.00 €
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
17922001
View Documents
Invitrogen™
416-5743-80
25 μg
188.00 €
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

OVA257-264 (SIINFEKL) peptide bound to H-2Kb Monoclonal antibody specifically detects OVA257-264 (SIINFEKL) peptide bound to H-2Kb in Mouse samples. It is validated for Flow Cytometry
Specifications

Specifications

OVA257-264 (SIINFEKL) peptide bound to H-2Kb
0.2 mg/mL
Monoclonal
Liquid
RUO
PBS with BSA and 0.09% sodium azide; pH 7.2
IgG1 κ
Affinity chromatography
eBio25-D1.16 (25-D1.16)
Flow Cytometry
Brilliant Violet 650
Mouse
Mouse
H-2Kb-SIINFEKL; OVA257-264; OVA257-264 H-2Kb; OVA-Kb
Primary
4°C, store in dark, DO NOT FREEZE!
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.