Learn More
Caframo™ Overhead Stirrer Clamp
Overhead Stirrer Clamp designed to accommodate stirrer support rods up to 16 mm (5/8 in.) diameter. Attaches overhead stirrer to Caframo and Fisherbrand stands (14-500-354 and 14-500-355).
Brand:  Caframo™ A122
Product Code. 16853953
  • 138.00 € / Each
Estimated Shipment: 13-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • This clamp is an alternative to the A120 and A120FSR (14-500-6, 14-500-357)
  • Compatible with larger diameter support rods, up to 16 mm (5/8 in.)
Specifications
Specifications
Caframo™ and Fisherbrand™ Stands (14-500-354 and 14-500-355)
81 mm
40 mm
Key Only
Clamp
Cast Zinc, Aluminum
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Caframo™ Overhead Stirrer Clamp > Overhead Stirrer Clamp; Material: 420 Stainless Steel