missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eagle Overpacks with Screw-On Lid
Eagle Overpacks are ideal for safe storage, handling and transportation of hazardous materials, spill containment and clean-up of hazardous materials, original shipping container, bio waste disposal and incineration.
433.00 € - 481.00 €
Specifications
Color | Yellow |
---|---|
For Use With (Application) | Safe Storage, Handling and Transportation of Hazardous Materials |
Material | Polyethylene |
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
17772308
|
Eagle
EDR16610YL |
247 L |
433.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17782308
|
Eagle
EDR16900YL |
361 L |
481.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Eagle Overpack Poly Drums are constructed of yellow blow-moulded high-density polyethylene (HDPE) with UV inhibitors, is durable and lightweight, weather and chemical resistant, and has a yellow screw-on lid
- Ideal for safe storage, handling and transportation of hazardous materials, spill containment and cleanup of hazardous materials, original shipping container, bio waste disposal and incineration
- Overpacks - for use with larger non-damaged containers, can and drums of non-leaking hazardous materials - if leaking, must pack with sufficient sorbents for - NO FREE LIQUIDS
Specifications
Yellow | |
Safe Storage, Handling and Transportation of Hazardous Materials | |
Polyethylene |