missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Owl™ A1 Large Gel Electrophoresis System Standard Comb

Eliminate comb assembly with this assortment of heavy-duty, one-piece combs. For use with the Fisherbrand™ Owl™ A1 Large Gel Electrophoresis Systems.

56.10 € - 78.55 €

Specifications

For Use With (Equipment) Owl A1 Large Gel Electrophoresis System
Product Line Owl
Type Comb
View More Specs

Products 4
Product Code Brand Description Teeth Count Thickness (Metric) Teeth Price Quantity & Availability  
Product Code Brand Description Teeth Count Thickness (Metric) Teeth Price Quantity & Availability  
12473647
View Documents
Thermo Scientific™
A1-RL-25C
Rapid load comb 25 (2X) 1.0 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11986916
View Documents
Thermo Scientific™
A1-RL-25D
Rapid load comb 25 (2X) 1.5 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
12483647
View Documents
Thermo Scientific™
A1-RL-9C
Rapid load comb 9 (1X) 1.0 mm
56.10 €
Each
Estimated Shipment: 10-02-2025
to see stock.
Add to basket
 
12493647
View Documents
Thermo Scientific™
A1-RL-9D
Rapid load comb 9 (1X) 1.5 mm
56.10 €
Each
Estimated Shipment: 10-02-2025
to see stock.
Add to basket
 
Description

Description

Eliminate comb assembly with this assortment of heavy-duty, one-piece Thermo Scientific™ Owl™ A1 Large Gel Electrophoresis System Combs, for use with Owl A1 Large Gel Electrophoresis Systems.

Heavy-duty, one-piece combs eliminate comb assembly.
Specifications

Specifications

Owl A1 Large Gel Electrophoresis System
Owl
Comb
Videos
SDS
Documents

Documents

Product Certifications