missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Owl™ A2 Large Gel Systems

Handle high throughput with consistent, reliable results with Thermo Scientific™ Owl™ A2 Large Gel Systems, ideal for detailed RNA/DNA analysis.

613.74 € - 1365.00 €

Specifications

Certifications/Compliance CE marked
Product Line Owl
Sample Size 8 to 432 (16-, 24- and 36-well combs)
Volume (Metric) Buffer 2.3 L
Dimensions (L x W x H) 14.8 x 12.8 x 4.1 in. (37.5 x 32.5 x 10.5 cm)
View More Specs

Products 3
Product Code Brand Type Price Quantity & Availability  
Product Code Brand Type Price Quantity & Availability  
11520255
View accessory products View Documents
Thermo Scientific™
A2
Large Gel Electrophoresis System N/A N/A N/A
11797105
View accessory products View Documents
Thermo Scientific™
A2
Large Gel Electrophoresis System N/A N/A N/A
11902915
View accessory products View Documents
Thermo Scientific™
A2-BP
Large, w/Buffer Exchange Ports
1365.00 €
Each
Estimated Shipment: 07-05-2025
to see stock.
Add to basket
 
Description

Description

Handle high throughput with consistent, reliable results with Thermo Scientific™Owl™ A2 Large Gel Systems, ideal for detailed RNA/DNA analysis.

Features:

  • Gel can be cast in varying lengths to help conserve agarose when fewer samples need to be run
  • Optional built-in buffer exchange ports allow for buffer recirculation on extended runs
  • UVT gel tray has gasketed end gates for leakproof seal without tape
  • Built-in leveling ensures flat, uniformly thick gels
  • Comb slots allow two, three or four sample sets of equal lengths to be run on the same gel
  • Microwell-format combs provide fast, error-free sample loading with a multichannel pipetter
  • Samples may be loaded directly from a 96-well plate, 8 or 12 at a time

Includes:

  • Buffer chamber with three-point leveling base
  • Thermo Scientific™ SuperSafe™ lid with attached power supply leads
  • UVT gel tray
  • Gasketed end gates
  • Three combs (16-, 24- and 36-well), 1.5mm thick
Specifications

Specifications

CE marked
8 to 432 (16-, 24- and 36-well combs)
14.8 x 12.8 x 4.1 in. (37.5 x 32.5 x 10.5 cm)
37.5 cm
12.8 in.
20 cm
10.5 cm
Owl
2.3 L
14.8 in.
25 cm
32.5 cm
4.1 in.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Owl™ A2 Large Gel Systems

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.