Learn More
Thermo Scientific™ Owl™ A3-1 Large-Gel Electrophoresis System
Conduct plant genomics studies with Thermo Scientific™ Owl™ A3-1 Large-Gel Electrophoresis System.
Brand: Thermo Scientific™ A3-1
Code : 04
Additional Details : Weight : 8.00000kg
Description
Conduct plant genomics studies with Thermo Scientific™ Owl™ A3-1 Large-Gel Electrophoresis System.
Features:- Horizontal system can run from 25 to 600 samples on one gel
- Gel can be cast in varying lengths to help conserve agarose when fewer samples need to be run
- Built-in buffer exchange ports allow for buffer recirculation on extended runs
- UVT gel tray has gasketed end gates for leakproof seal without tape
- Built-in leveling ensures flat, uniformly thick gels
- Large number of comb slots provides sample number and resolving distance flexibility
- Microwell-format combs provide fast, error-free sample loading with a multichannel pipetter
- Samples may be loaded directly from a 96-well plate, 8 or 12 at a time
Ordrring Information:
Customized combs are available (Order SKU # XCM). Please contact your Customer Service or Sales Representative for further details.
Includes:
Buffer chamber with three-point leveling base, SuperSafe™ lid with attached power supply leads, UVT gel tray, four combs (two 25- and two 50-well), built-in buffer exchange ports and bubble level
Specifications
CE marked | |
23 x 40 cm | |
Large Gel Electrophoresis System | |
4.5 L | |
20.63 in. | |
40 cm | |
32 cm | |
4.25 in. | |
250 V |
Gel size: 23 x 40cm; Buffer volume: 4.5L; L x W x H: 53 x 32 x 11cm | |
Owl | |
25 to 600 | |
20.63 x 12.5 x 4.25 in. (53 x 32 x 11 cm) | |
53 cm | |
12.5 in. | |
23 cm | |
11 cm |