missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Owl™ Aluminum-Backed Sequencers

Easily set up applications with even heat distribution for smile-free results with manual Thermo Scientific™ Owl™ Aluminum-Backed Sequencers.

Brand:  Thermo Scientific™ S4S

 View more versions of this product

Product Code. 11923075

  • 2235.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Easily set up applications with even heat distribution for smile-free results with manual Thermo Scientific™ Owl™ Aluminum-Backed Sequencers.

Features:

  • Floating aluminum heat sink reduces chance of plates cracking during runs.
  • Upper buffer chamber (UBC) requires minimal buffer and is easily drained via inline drain port into removable bottle.
  • Lower buffer chamber (LBC) is removable for simple buffer disposal.
  • Side clamps press gel plates against gasket seated on UBC, applying even pressure across plate and forming a leak-proof seal resulting in flat, even banding patterns.
  • Clamps slide over glass plates for easy setup and removal of gels.

Includes:

  • Removable LBC
  • Heat sink
  • Two side clamps
  • Two blank glass plates
  • Two notched glass plates
  • Two sharktooth combs (78 teeth, 0.4mm thick)
  • Well comb (40 well, 0.4mm thick)
  • Two spacer sets (0.4mm)
  • Adhesive temperature indicator
  • UBC with removable drain bottle (with safety interlocking lid and attached power supply lead)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CE marked
10.5 x 9 x 19.5 in. (27 x 23 x 50cm)
19.5 in.
10.5 in.
27 cm
27 fl oz.
9 in.
23 cm
Gel: 20 x 45cm; 950mL; 27 x 23 x 50cm
40-well
50 cm
17.716 in.
45 cm
800mL
7.87 in.
20 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.