missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Owl™ D2 Wide-Gel Electrophoresis System

Run 10 to 80 samples on one gel with this wide-gel electrophoresis system.

Brand:  Thermo Scientific™ D2-BP

 View more versions of this product

Product Code. 12404607

  • 1175.00 € / Each
Estimated Shipment: 29-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Run 10 to 80 samples on one gel with the Thermo Scientific™ Owl™ D2 Wide-Gel Electrophoresis System.

  • Wide gel size allows 15 or 30 samples to be loaded per row with a micro well comb.
  • Using special combs and a multichannel pipetter, samples may be loaded directly from a 96-well plate, eight or 12 at a time.
  • System is excellent for screening PCR samples or routine DNA fragment analysis.
  • Small footprint saves space on lab bench.
  • Incorporates Thermo Scientific™ EasyCast™ casting design with external caster—gel tray is fitted with gasket, allowing it to fit snugly into caster and providing a leakproof seal without tape.
  • Electrode placement allows gel to be run quickly without compromising resolution.

Includes:

  • Buffer chamber
  • Thermo Scientific™ SuperSafe™ lid with attached power supply leads
  • EasyCast gasketed UVT gel tray
  • External multiple gel caster
  • Four combs (two 30-well and two 40-well, 1.5mm thick)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CE marked
Owl
10 to 80
7.38 x 7.75 x 4 in. (19 x 20 x 10 cm)
19 cm
7.75 in.
20 cm
4 in.
150 V
Complete System with Buffer Exchange Ports
Wide w/Buffer Exchange Ports
600 mL
7.38 in.
14.4 cm
6.5
10.2 cm
10 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.