missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Owl™ Dual-Gel Vertical Electrophoresis Systems
Quickly and easily cast and run one or two gels in the same device with these Thermo Scientific™ Owl™ Dual-Gel Vertical Electrophoresis Systems.
1090.00 € - 1900.00 €
Product Code | Brand | Description | Volume (Metric) Buffer | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Volume (Metric) Buffer | Price | Quantity & Availability | ||||
11903015
|
Thermo Scientific™
P10DS-1 |
P10DS Complete System plus Caster | 1250 mL | ||||||
11913015
|
Thermo Scientific™
P10DS-2 |
P10DS Complete System | 1250 mL |
1620.00 €
Each |
Estimated Shipment: 29-08-2025
Log in to see stock. |
||||
12474677
|
Thermo Scientific™
P10DS-3 |
P10DS Apparatus Only | 1250 mL |
1470.00 €
Each |
Estimated Shipment: 11-12-2024
Log in to see stock. |
||||
11913005
|
Thermo Scientific™
P8DS-1 |
P8DS Complete System plus Caster | 300 mL |
1460.00 €
Each |
Estimated Shipment: 16-12-2024
Log in to see stock. |
||||
11923005
|
Thermo Scientific™
P8DS-2 |
P8DS Complete System | 300 mL |
1090.00 €
Each |
Estimated Shipment: 09-12-2024
Log in to see stock. |
||||
12404707
|
Thermo Scientific™
P8DS-3 |
P8DS Apparatus Only | 300 mL |
1315.00 €
Each |
Estimated Shipment: 18-12-2024
Log in to see stock. |
||||
11973005
|
Thermo Scientific™
P9DS-1 |
P9DS Complete System plus Caster | 550 mL |
1380.00 €
Each |
Estimated Shipment: 23-01-2025
Log in to see stock. |
||||
11983005
|
Thermo Scientific™
P9DS-2 |
P9DS Complete System | 550 mL |
1370.00 €
Each |
Estimated Shipment: 16-12-2024
Log in to see stock. |
||||
11993005
|
Thermo Scientific™
P9DS-3 |
P9DS Apparatus Only | 550 mL |
1180.00 €
Each |
Estimated Shipment: 02-12-2024
Log in to see stock. |
||||
Descripción
Quickly and easily cast and run one or two gels in the same device using Thermo Scientific™ Owl™ Dual-Gel Vertical Electrophoresis Systems—comprised of the P8DS, P9DS and P10DS Dual Gel Systems.
Simple, Six-Step Use:
- Secure the glass cassettes
- Pour the agarose
- Plug in the specially designed trough
- Cast polyacrylamide solution in the cassettes
- Load samples
- Run the gel
Features:
- Manufactured using a time-tested acrylic fabrication technology, providing a durable, long-lasting system
- Central cooling core provides even cooling across the gel surface for outstanding resolution and clear banding
- Cooling is achieved by circulating fluid through the core with external circulator or cool tap water
- Very simple clamping mechanism makes loading glass cassettes and most manufacturers' precast gels trouble free
Recommended for:
- SDS electrophoresis
- The second dimension of two-dimensional (2-D) electrophoresis
- Agarose electrophoresis
- Native electrophoresis
Especificaciones
Owl |