Learn More
Thermo Scientific™ Owl™ EasyCast™ B1 Mini Gel Electrophoresis Systems
Cast and run a gel in the same chamber with no tape or additional parts required with the leakproof casting of mini gel electrophoresis systems.
Brand:  Thermo Scientific™ B1-BP
 View more versions of this product
Product Code. 11942935
  • 740.00 € / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
Cast and run a gel in the same chamber with no tape or additional parts required with the leakproof casting of Thermo Scientific™ Owl™ EasyCast™ B1 Mini Gel Electrophoresis Systems. Durability and ease of use makes this the perfect choice for students and high volume researchers.
Features:
  • System offers a wide range of combs.
  • Two comb slots on gel tray double sample capacity by doubling number of sample wells.
  • UVT gel trays are silk screened with a florescent ruler for easy measurement of bands.
Includes:
  • Main buffer chamber
  • Thermo Scientific™ SuperSafe™ lid with attached power supply leads
  • Thermo Scientific™ EasyCast™ Gasketed UVT gel tray
  • Two Combs: 10 & 14 Well, Double-sided, 1.0/1.5mm thick
Ordering Information:
Available with optional buffer exchange ports, (B1-BP), for use with an external recirculation pump.
Warranty and Service Offering:
  • Three years
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CE marked
Mini
Mini Gel Electrophoresis System
600 mL
8.63 in.
11 cm
15 cm
3.75 in.
EasyCast w/buffer exchange ports
Owl
5 to 34
8.63 x 5.88 x 3.75 in. (22 x 15 x 9.5 cm)
22 cm
5.88 in.
9 cm
9.5 cm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products