missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Owl™ EasyCast™ B2 Mini Gel Electrophoresis Systems
Achieve superior flexibility in a small footprint with our B2 mini gel electrophoresis systems.
663.00 € - 745.00 €
Specifications
Certifications/Compliance | CE marked |
---|---|
Gel Size | Mini |
Product Line | Owl |
Type | Mini Gel Electrophoresis System |
Sample Size | 8 to 48 |
Product Code | Brand | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
11717185
|
Thermo Scientific™
B2-BP |
EasyCast w/buffer exchange ports |
745.00 €
Each |
In Stock
Log in to see stock. |
|||||
11972955
|
Thermo Scientific™
B2 |
EasyCast |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Achieve superior flexibility in a small footprint with Thermo Scientific™ Owl™ EasyCast™ B2 Mini Gel Electrophoresis Systems. The simple-to-use casting method, as well as multiple comb and tray options, make this product Owl's most popular system.
Features:
- Two comb slots on the U.V. Transmissible (UVT) gel tray double sample capacity by doubling the number of sample wells
- UVT gel trays are silk screened with a florescent ruler for easy measurement of bands
Ordering Information:
Available with optional buffer exchange ports (B2-BP) for use with an external recirculation pump
Includes:
- Buffer chamber
- Thermo Scientific™ SuperSafe™ lid with attached power supply leads
- EasyCast gasketed U.V. Transmissible (UVT) gel tray
- Two combs: 12 & 20 well, double-sided, 1.0/1.5 mm thick
Specifications
CE marked | |
Owl | |
8 to 48 | |
9.63 x 7.06 x 3.75 in. (24.5 x 18 x 9.5 cm) | |
24.5 cm | |
7.06 in. | |
12 cm | |
9.5 cm |
Mini | |
Mini Gel Electrophoresis System | |
800 mL | |
9.63 in. | |
14 cm | |
18 cm | |
3.75 in. |